Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC2, Mouse, Clone: M2II-12, Abnova™

Catalog No. 89111386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-386 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-386 Supplier Abnova Corporation Supplier No. MAB3383
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ABCC2.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein is expressed in the canalicular (apical) part of the hepatocyte and functions in biliary transport. Substrates include anticancer drugs such as vinblastine; therefore, this protein appears to contribute to drug resistance in mammalian cells. Several different mutations in this gene have been observed in patients with Dubin-Johnson syndrome (DJS), an autosomal recessive disorder characterized by conjugated hyperbilirubinemia. [provided by RefSeq

Specifications

Antigen ABCC2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone M2II-12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ABCC2.
Dilution Western Blot (1:20-50) Immunohistochemistry (Frozen sections) (1:20) Immunocytochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In serum-free cell culture supernatant (0.09% sodium azide, 0.7% BSA)
Gene ABCC2
Gene Alias ABC30/CMOAT/DJS/KIAA1010/MRP2/cMRP
Gene Symbols ABCC2
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to amino acids 860-950 of human ABCC2.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1244
Test Specificity Reacts with an internal epitope of human cMOAT/MRP2. Does not cross-react with human MDR1, MRP1, MRP3, and MRP5 gene products.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.