Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcc2, Mouse, Clone: M2III-5, Abnova™

Catalog No. 89111394 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-394 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-394 Supplier Abnova Corporation Supplier No. MAB3399
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Abcc2.

Specifications

Antigen Abcc2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone M2III-5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Abcc2.
Dilution Immunocytochemistry (1:20-1:50) Immunohistochemistry (Frozen sections) (1:20) Western Blot (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (0.09% sodium azide)
Gene Abcc2
Gene Alias Cmoat/Mrp2
Gene Symbols Abcc2
Host Species Mouse
Immunogen Recombinant protein corresponding to chimeric protein with 202 amino acids at the C-terminus of rat Abcc2 and bacterial maltose-binding protein.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25303
Test Specificity M2III-5 reacts with an internal epitope of MRP2 (ABCC2), a 190-200 KDa transmembrane protein. M2III-5 does not cross-react with the human MDR1 P-gp, MRP1, MRP3 or MRP5 gene products.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.