Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADS, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104029 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-029 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-029 Supplier Abnova Corporation Supplier No. H00000035A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ACADS.

This gene encodes a a tetrameric mitochondrial flavoprotein, which is a member of the acyl-CoA dehydrogenase family. This enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Mutations in this gene have been associated with Short Chain Acyl-CoA Dehydrogenase Deficiency. [provided by RefSeq

Sequence: SKEQKQAWVTPFTSGDKIGCFALSEPGNGSDAGAASTTARAEGDSWVLNGTKAWITNAWEASAAVVFASTDRALQNKGISAFLVPMPTPG

Specifications

Antigen ACADS
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ACADS.
Formulation 50% glycerol
Gene ACADS
Gene Accession No. NM_000017
Gene Alias ACAD3/SCAD
Gene Symbols ACADS
Host Species Mouse
Immunogen ACADS (NP_000008, 132 a.a. to 221 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 35
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.