Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAN, Mouse, Clone: 14A6 3B2, Abnova™

Catalog No. 89111524 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-524 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-524 Supplier Abnova Corporation Supplier No. MAB3670
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant ACAN.

This gene is a member of the aggrecan/versican proteoglycan family. The encoded protein is an integral part of the extracellular matrix in cartilagenous tissue and it withstands compression in cartilage. Mutations in this gene may be involved in skeletal dysplasia and spinal degeneration. Multiple alternatively spliced transcript variants that encode different protein isoforms have been observed in this gene. [provided by RefSeq

Specifications

Antigen ACAN
Applications ELISA
Classification Monoclonal
Clone 14A6 3B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant ACAN.
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS, pH 8.0
Gene ACAN
Gene Alias AGC1/AGCAN/CSPG1/CSPGCP/MSK16/SEDK
Gene Symbols ACAN
Host Species Mouse
Immunogen Recombinant protein corresponding to human ACAN.
Purification Method Protein A purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 176
Test Specificity This antibody recognizes natural and recombinant human ACAN.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.