Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT1, Mouse, Clone: AT15E5, Abnova™

Catalog No. 89109375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-375 Supplier Abnova Corporation Supplier No. MAB11206
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ACAT1.

This gene encodes a mitochondrially localized enzyme that catalyzes the reversible formation of acetoacetyl-CoA from two molecules of acetyl-CoA. Defects in this gene are associated with 3-ketothiolase deficiency, an inborn error of isoleucine catabolism characterized by urinary excretion of 2-methyl-3-hydroxybutyric acid, 2-methylacetoacetic acid, tiglylglycine, and butanone. [provided by RefSeq

Specifications

Antigen ACAT1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT15E5
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ACAT1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.01% Sodium Azide.
Gene ACAT1
Gene Alias ACAT/MAT/T2/THIL
Gene Symbols ACAT1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 34-427 of human ACAT1.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 38
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.