Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-ACD, Clone: 1G10.3,

Catalog No. MABE903MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE903MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE903MI Supplier MilliporeSigma™ Supplier No. MABE903
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects ACD clone: 1G10.3 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

Adrenocortical dysplasia protein homolog (also known as POT1 and TIN2 organizing protein or TIN2-interacting protein) is a nuclear protein encoded by the ACD gene in human (also known as PIP1, PTOP, TINT1, TPP1). It a component of the shelterin complex (telosome) and is involved in the regulation of telomere length and protection. Shelterin associates with arrays of double-stranded TTAGGG repeats added by telomerase and protects chromosome ends. Without its protective activity, telomeres are no longer hidden from the DNA damage surveillance, and chromosome ends are inappropriately processed by DNA repair pathways. The ACD-POT1 heterodimer is reported to enhance telomere elongation by increasing telomerase processivity (PMID 17237768).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACD
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1G10.3
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96AP0
Gene Symbols ACD; PIP1; PTOP; TINT1; TPP1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human ACD.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001075955
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.