Learn More
ACE, Mouse, Clone: 3C5, Abnova™
Mouse monoclonal antibody raised against ACE.
Supplier: Abnova Corporation MAB5228
Description
This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes - the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined. [provided by RefSeq
Specifications
ACE | |
Monoclonal | |
Unconjugated | |
In tissue culture supernatant (0.09% sodium azide) | |
ACE1, CD143, DCP, DCP1, MGC26566 | |
Mouse | |
1 mL | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
3C5 | |
angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 | |
ACE | |
ACE | |
Denatured human ACE. | |
RUO | |
1636 | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.