Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acidic Cytokeratin, Mouse, Clone: SPM115, Abnova™

Catalog No. 89110218 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-218 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-218 Supplier Abnova Corporation Supplier No. MAB12076
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Acidic Cytokeratin.

Specifications

Antigen Acidic Cytokeratin
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SPM115
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Acidic Cytokeratin.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Alias CK10/K10/KPP
Gene Symbols KRT10
Host Species Mouse
Immunogen Solubilized keratin extract from human stratum corneum.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3858/3861/3866/3868/3880
Test Specificity This monoclonal antibody recognizes KRT10 (56.5kDa), KRT14 (50kDa), KRT15 (50kDa), KRT16 (48kDa), and KRT19 (40kDa).
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.