Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT11, Mouse, Clone: J4B2, Abnova™

Catalog No. 89110614 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-614 Supplier Abnova Corporation Supplier No. MAB1770
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ACOT11.

This gene encodes a protein with acyl-CoA thioesterase activity towards medium (C12) and long-chain (C18) fatty acyl-CoA substrates. Expression of a similar murine protein in brown adipose tissue is induced by cold exposure and repressed by warmth. Expression of the mouse protein has been associated with obesity, with higher expression found in obesity-resistant mice compared with obesity-prone mice. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen ACOT11
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone J4B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ACOT11.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ACOT11
Gene Alias BFIT/BFIT1/BFIT2/KIAA0707/STARD14/THEA/THEM1
Gene Symbols ACOT11
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 19-250 of human ACOT11.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26027
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.