Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT7, Mouse, Clone: AT1D5, Abnova™

Catalog No. 89109381 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-381 Supplier Abnova Corporation Supplier No. MAB11211
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ACOT7.

This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized. [provided by RefSeq

Specifications

Antigen ACOT7
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1D5
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ACOT7.
Dilution Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.02% Sodium Azide.
Gene ACOT7
Gene Alias ACH1/ACT/BACH/CTE-II/LACH/LACH1/MGC1126/hBACH
Gene Symbols ACOT7
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-370 of human ACOT7.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11332
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.