Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACPP, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104033 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-033 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-033 Supplier Abnova Corporation Supplier No. H00000055A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ACPP.

This gene encodes an enzyme that catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland. An alternatively spliced transcript variant encoding a longer isoform has been found for this gene. This isoform contains a transmembrane domain and is localized in the plasma membrane-endosomal-lysosomal pathway. [provided by RefSeq

Sequence: YASCHLTELYFEKGEYFVEMYYRNETQHEPYPLMLPGCSPSCPLERFAELAGPVIPQDWSTECMTTNSHQVLKVIFAVAFCLISAVLMVLLFIHIRRGLCWQRESYGNI

Specifications

Antigen ACPP
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ACPP.
Formulation 50% glycerol
Gene ACPP
Gene Accession No. BC007460
Gene Alias ACP-3/ACP3/PAP
Gene Symbols ACPP
Host Species Mouse
Immunogen ACPP (AAH07460, 310 a.a. to 418 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.