Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACR, Mouse, Clone: ACR-2, Abnova™

Catalog No. 89111492 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-492 Supplier Abnova Corporation Supplier No. MAB3593
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ACR.

Specifications

Antigen ACR
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone ACR-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ACR.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ACR
Gene Symbols ACR
Host Species Mouse
Immunogen Native purified porcine ACR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397098
Test Specificity This antibody reacts with various forms of porcine acrosin (55, 53, 45 and 35 KDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa.
Target Species Pig
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.