Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACRV1, Mouse, Clone: Hs-14, Abnova™

Catalog No. 89111615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-615 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-615 Supplier Abnova Corporation Supplier No. MAB3824
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native purified ACRV1.

The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation. [provided by RefSeq

Specifications

Antigen ACRV1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone Hs-14
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native purified ACRV1.
Dilution Immunocytochemistry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene VCP
Gene Alias IBMPFD/MGC131997/MGC148092/MGC8560/TERA/p97
Gene Symbols VCP
Host Species Mouse
Immunogen Native purified human VCP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7415
Test Specificity The antibody Hs-14 reacts with VCP (valosin-containing protein) a 220 kDa protein previously identified under the general name “intra-acrosomal protein”.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.