Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDHS, Mouse, Clone: Hs-8, Abnova™

Catalog No. 89108247 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-247 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-247 Supplier Abnova Corporation Supplier No. MAB0919
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ACRV1.

This gene encodes a protein belonging to the glyceraldehyde-3-phosphate dehydrogenase family of enzymes that play an important role in carbohydrate metabolism. Like its somatic cell counterpart, this sperm-specific enzyme functions in a nicotinamide adenine dinucleotide-dependent manner to remove hydrogen and add phosphate to glyceraldehyde 3-phosphate to form 1,3-diphosphoglycerate. During spermiogenesis, this enzyme may play an important role in regulating the switch between different energy-producing pathways, and it is required for sperm motility and male fertility. [provided by RefSeq

Specifications

Antigen GAPDHS
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone Hs-8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ACRV1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene GAPDHS
Gene Alias GAPD2/GAPDH-2/GAPDS/HSD-35
Gene Symbols GAPDHS
Host Species Mouse
Immunogen Native purified human GAPDHS.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26330
Test Specificity The antibody Hs-8 reacts with GAPDHS, the sperm-specific glyceraldehyde phosphate dehydrogenase, previously identified under the general name “intra-acrosomal protein”.
Target Species Human, Pig
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.