Learn More
ACTA2, Mouse, Clone: IA4, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of ACTA2.
Supplier: Abnova Corporation MAB1492
Description
The protein encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Defects in this gene cause aortic aneurysm familial thoracic type 6. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Specifications
| ACTA2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (2 ug/mL) Immunohistochemistry (Frozen sections) (0.3-1.2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ACTA2 | |
| ACTA2 | |
| A synthetic peptide corresponding to N-terminus of human ACTA2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| IA4 | |
| Mouse monoclonal antibody raised against synthetic peptide of ACTA2. | |
| Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
| AAT6/ACTSA | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 59 | |
| Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.