Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTB, Mouse, Clone: BA3R, Abnova™

Catalog No. 89136766 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-766 Supplier Abnova Corporation Supplier No. MAB9743
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of ACTB.

Specifications

Antigen ACTB
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone BA3R
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of ACTB.
Dilution Western Blot (1:1000-1:10000) Immunohistochemistry (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.05% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of ACTB.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes native and denatured forms of beta-Actin (∼42 KDa).
Target Species Chicken, Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.