Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTB, Mouse, Clone: G043, Abnova™

Catalog No. 89136430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-430 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-430 Supplier Abnova Corporation Supplier No. MAB8182
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of ACTB.

Specifications

Antigen ACTB
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone G043
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of ACTB.
Dilution Western Blot (1:1000-1:4000) Immunofluorescence (1:100-1:400) The optimal working dilution should be determined by the end user.
Formulation In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of ACTB.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Canine, Carp, Chicken, Feline, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.