Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Actin, Clone: C4, Alexa Fluor 555,

Catalog No. MAB1501AF55 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB1501AF55 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1501AF55 Supplier MilliporeSigma™ Supplier No. MAB1501AF555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Actin clone: C4 in Human samples, and it is validated for Immunocytochemistry

Actin is a multi-functional, highly conserved cytoskeletal protein that is found in most eukaryotic cells. This 42kDa protein assumes two physical forms: globular actin (G-actin), which may serve as a cytoplasmic storage pool, and fibrous actin (F-actin), which, in conjunction with myosin, generates muscle contraction. In non-muscle cells, actin appears to be involved in a variety of functions, such as cell motility, exocytosis, and phagocytosis (True, 1990). Distribution of the six known isoforms of actin - four muscle actins (alpha-skeletal, alpha-vascular smooth, alpha-cardiac, and gamma-enteric smooth) and two cytoplasmic actins (alpha and gamma) is tissue specific.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunocytochemistry
Classification Monoclonal
Clone C4
Conjugate Alexa Fluor 555
Formulation Purified mouse antibody conjugate in buffer containing PBS, 15mg/mL BSA, and 0.1% sodium azide.
Gene Accession No. P68139
Gene Symbols ACTA1; ACTA
Host Species Mouse
Immunogen Purified chicken gizzard actin (Lessard, 1988).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001026234.1
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.