Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Active-β-Catenin, Clone: 8E7, Alexa Fluor 488,

Catalog No. 05665AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-665-AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-665-AF488 Supplier MilliporeSigma™ Supplier No. 05665AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Active-β-Catenin clone: 8E7 in Human, Mouse samples, and it is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Catenin beta-1 (UniProt P35222; also known as beta-Catenin) is encoded by the CTNNB1 (also known as CTNNB, MRD19) gene (ORF OK/SW-cl.35, PRO2286; Gene ID 1499) in human. beta-Catenin is a multifunctional protein involved in the regulation of cell–cell adhesion and in the control of the Wnt signaling pathway. beta-Catenin contains in its central region 12 armadillo repeats that mediate its interaction with a variety of proteins including axin, adenomatous polyposis coli (APC), and T cell factor (TCF) family transcription factors. The C-terminal domain (CTD) of beta-catenin is implicated in transcriptional functions, while the N-terminal domain (NTD) contains several posttranslational modification (ubiquitination, acetylation, and phosphorylation) sites, including the Ser33/Ser37/Thr41 residues, important for beta-catenin stability regulation. In the absence of Wnt pathway activation, beta-catenin is retained in the cytoplasm in a 'destruction complex' with Axin, APC, and GSK3beta, where it is constitutively destined for proteasomal degradation as a result of constant phosphorylation by GSK3beta. Activation of the Wnt pathway results in stabilization and nuclear translocation of beta-catenin, resulting in beta-catenin target genes transcription. In addition to GSK3beta, PKCalpha is also reported to phosphorylate the Ser33/37/Thr41 residues of beta-catenin, and beta-catenin Ser45 is a known phosphorylation site for CK1alpha. In addition, HDAC6 and p300/CBP-associated factor (PCAF) regulate beta-catenin Lys49 acetylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Active-β-Catenin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 8E7
Conjugate Alexa Fluor 488
Formulation Purified mouse conjugate antibody in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. P35222
Gene Symbols CTNNB1; CTNNB; PRO2286
Host Species Mouse
Immunogen Fusion protein corresponding to residues 1-100 of human β-catenin with a c-terminal histidine tag.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001895
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.