Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM29, Mouse, Clone: 1F6, Abnova™

Catalog No. 89106458 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-458 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-458 Supplier Abnova Corporation Supplier No. H00011086M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADAM29.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is highly expressed in testis and may be involved in human spermatogenesis. Alternative splicing results in multiple transcript variants that encode the same protein. [provided by RefSeq

Sequence: GMNHDEDTCRCSQPRCIMHEGNPPITKFSNCSYGDFWEYTVERTKCLLETVHTKDIFNVK

Specifications

Antigen ADAM29
Applications ELISA
Classification Monoclonal
Clone 1F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADAM29.
Formulation ascites with no preservative
Gene ADAM29
Gene Accession No. NM_014269
Gene Alias svph1
Gene Symbols ADAM29
Host Species Mouse
Immunogen ADAM29 (NP_055084,339 a.a. ∽ 398 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11086
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.