Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS15, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-681 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-681 Supplier Abnova Corporation Supplier No. H00170689A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ADAMTS15.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the proteins encoded by ADAMTS1 and ADAMTS8. [provided by RefSeq

Sequence: AVDCRGSAGQRTVPACDAAHRPVETQACGEPCPTWELSAWSPCSKSCGRGFQRRSLKCVGHGGRLLARDQCNLHRKPQELDFCVLRPC

Specifications

Antigen ADAMTS15
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ADAMTS15.
Formulation 50% glycerol
Gene ADAMTS15
Gene Accession No. NM_139055
Gene Alias MGC126403
Gene Symbols ADAMTS15
Host Species Mouse
Immunogen ADAMTS15 (NP_620686, 863 a.a. to 950 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170689
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.