Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-474 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-474 Supplier Abnova Corporation Supplier No. H00011173A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ADAMTS7.

The protein encoded by this gene is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) family. Members of this family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two C-terminal TS motifs. [provided by RefSeq

Sequence: VQRRLVKCVNTQTGLPEEDSDQCGHEAWPESSRPCGTEDCEPVEPPRCERDRLSFGFCETLRLLGRCQLPTIRTQCCRSCSPPSHGAPSRGHQRVARR

Specifications

Antigen ADAMTS7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ADAMTS7.
Formulation 50% glycerol
Gene ADAMTS7
Gene Accession No. NM_014272
Gene Alias ADAM-TS7/DKFZp434H204
Gene Symbols ADAMTS7
Host Species Mouse
Immunogen ADAMTS7 (NP_055087, 1589 a.a. to 1686 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11173
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.