Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104044 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-044 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-044 Supplier Abnova Corporation Supplier No. H00000113A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ADCY7.

This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences. [provided by RefSeq

Sequence: HKHQMQDASRDLFTYTVKCIQIRRKLRIEKRQQENLLLSVLPAHISMGMKLAIIERLKEHGDRRCMPDNNFHSLYVKRHQN

Specifications

Antigen ADCY7
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ADCY7.
Formulation 50% glycerol
Gene ADCY7
Gene Accession No. NM_001114
Gene Alias AC7/FLJ36387/KIAA0037
Gene Symbols ADCY7
Host Species Mouse
Immunogen ADCY7 (NP_001105, 197 a.a. to 277 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.