Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus, Mouse, FITC, Clone: 143, Abnova™

Catalog No. 89135976 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-976 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-976 Supplier Abnova Corporation Supplier No. MAB5460
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against adenovirus.

Specifications

Antigen Adenovirus
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 143
Conjugate FITC
Description Mouse monoclonal antibody raised against adenovirus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Adenovirus type 6 infected cells.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the hexon group antigen of many Adenovirus serotypes. Known reactivity with 34 serotypes of Adenovirus including types 40 and 41 (40, 41, 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 14, 16, 18, 19, 20, 26, 31, 34, 35, 36 and 37). Does not react with Influenza A, Influenza B, RSV, Parainfluenza 1, 2 & 3, Mycoplasma pneumonia, H. pylori and Mammalian cells.
Target Species Adenovirus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.