Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus hexon, Mouse, Clone: AF-3H12, Abnova™

Catalog No. 89134565 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-565 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-565 Supplier Abnova Corporation Supplier No. MAB0016
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against adenovirus hexon.

Specifications

Antigen Adenovirus hexon
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AF-3H12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against adenovirus hexon.
Dilution Immunocytochemistry (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES/KOH, 150mM NaCl, pH 7.5 (50% glycerol)
Host Species Mouse
Immunogen Wild type adenovirus type 5 particles.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.