Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADH1A, Mouse, Clone: 1E5, Abnova™

Catalog No. 89107925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-925 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-925 Supplier Abnova Corporation Supplier No. MAB0349

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against native ADH1A.

This gene encodes class I alcohol dehydrogenase, alpha subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class I alcohol dehydrogenase, consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster. This gene is monomorphic and predominant in fetal and infant livers, whereas the genes encoding beta and gamma subunits are polymorphic and strongly expressed in adult livers. [provided by RefSeq

Specifications

Antigen ADH1A
Applications Immunocytochemistry
Classification Monoclonal
Clone 1.00E+05
Conjugate Unconjugated
Description alcohol dehydrogenase 1A (class I), alpha polypeptide
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ADH1A
Gene Alias ADH1
Gene Symbols ADH1A
Host Species Mouse
Immunogen Native purified human ADH1A.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.