Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin, Mouse, Clone: Adn279, Abnova™

Catalog No. 89136546 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-546 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-546 Supplier Abnova Corporation Supplier No. MAB8470
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native adiponectin.

Specifications

Antigen Adiponectin
Applications ELISA
Classification Monoclonal
Clone Adn279
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native adiponectin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified adiponectin from human serum.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recommended pairs for total adiponectin immunodetection in sandwich immunoassay (capture-detection) : Adn305-Adn279.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.