Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADIPOQ, Mouse, Clone: 236/1 GE9, Abnova™

Catalog No. 89111521 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-521 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-521 Supplier Abnova Corporation Supplier No. MAB3667
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant ADIPOQ.

This gene is expressed in adipose tissue exclusively. It encodes a protein with similarity to collagens X and VIII and complement factor C1q. The encoded protein circulates in the plasma and is involved with metabolic and hormonal processes. [provided by RefSeq

Specifications

Antigen ADIPOQ
Applications ELISA
Classification Monoclonal
Clone 236/1 GE9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant ADIPOQ.
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS
Gene ADIPOQ
Gene Alias ACDC/ACRP30/ADPN/APM-1/APM1/GBP28/adiponectin
Gene Symbols ADIPOQ
Host Species Mouse
Immunogen Recombinant protein corresponding to human ADIPOQ.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Test Specificity This antibody recognizes natural and recombinant human Adiponectin.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.