Learn More
ADK, Mouse, Clone: 7, Abnova™
Mouse monoclonal antibody raised against partial recombinant ADK.
Supplier: Abnova Corporation MAB9979
Description
This gene encodes adenosine kinase, an abundant enzyme in mammalian tissues. The enzyme catalyzes the transfer of the gamma-phosphate from ATP to adenosine, thereby serving as a regulator of concentrations of both extracellular adenosine and intracellular adenine nucleotides. Adenosine has widespread effects on the cardiovascular, nervous, respiratory, and immune systems and inhibitors of the enzyme could play an important pharmacological role in increasing intravascular adenosine concentrations and acting as anti-inflammatory agents. Alternative splicing results in two transcript variants encoding different isoforms. Both isoforms of the enzyme phosphorylate adenosine with identical kinetics and both require Mg2+ for activity. [provided by RefSeq]
Specifications
ADK | |
Monoclonal | |
Unconjugated | |
Western blot (1:500) The optimal working dilution should be determined by the end user. | |
ADK | |
ADK | |
Recombinant protein corresponding to amino acids 138-362 of human ADK. | |
100 μg | |
Primary | |
It can expression in HepG3 whole cell lysate. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
7 | |
Mouse monoclonal antibody raised against partial recombinant ADK. | |
In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
AK | |
Mouse | |
Affinity purification | |
RUO | |
132 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.