Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRBK1, Mouse, Clone: 5D5, Abnova™

Catalog No. 89110970 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-970 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-970 Supplier Abnova Corporation Supplier No. MAB2373
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant ADRBK1.

The product of this gene phosphorylates the beta-2-adrenergic receptor and appears to mediate agonist-specific desensitization observed at high agonist concentrations. This protein is an ubiquitous cytosolic enzyme that specifically phosphorylates the activated form of the beta-adrenergic and related G-protein-coupled receptors. Abnormal coupling of beta-adrenergic receptor to G protein is involved in the pathogenesis of the failing heart. [provided by RefSeq

Specifications

Antigen ADRBK1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant ADRBK1.
Dilution Immunohistochemistry (1:100) Western Blot (1:100) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant
Gene ADRBK1
Gene Alias BARK1/BETA-ARK1/FLJ16718/GRK2
Gene Symbols ADRBK1
Host Species Mouse
Immunogen Recombinant protein corresponding to human ADRBK1.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 156
Target Species Bovine, Human, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.