Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AEBP1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-049 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-049 Supplier Abnova Corporation Supplier No. H00000165A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant AEBP1.

The adipocyte enhancer binding protein 1 is a transcriptional repressor with carboxypeptidase (CP) activity. This protein binds to a regulatory sequence, adipocyte enhancer 1 (AE-1), located in the proximal promoter region of the adipose P2 (aP2) gene, which encodes the adipocyte fatty-acid binding protein. It is characterized as a member of the regulatory B-like CP family. This protein seems to be activated by a novel mechanism, whereby the direct binding of DNA enhances its protease activity. Adipocyte-enhancer binding protein 1 may play a role in differentiated vascular smooth muscle cells. [provided by RefSeq

Sequence: VTDEQGIPIANATISVSGINHGVKTASGGDYWRILNPGEYRVTAHAEGYTPSAKTCNVDYDIGATQCNFILARSNWKRIREIMAMNGNRPIPHIDPSRPMTP

Specifications

Antigen AEBP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant AEBP1.
Formulation 50% glycerol
Gene AEBP1
Gene Accession No. NM_001129
Gene Alias ACLP/FLJ33612
Gene Symbols AEBP1
Host Species Mouse
Immunogen AEBP1 (NP_001120, 912 a.a. to 1013 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.