Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin B1, Mouse, Clone: AFA-1, Abnova™

Catalog No. 89135608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-608 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-608 Supplier Abnova Corporation Supplier No. MAB4079

Mouse monoclonal antibody raised against Aflatoxin B1.

Specifications

Antigen Aflatoxin B1
Applications ELISA
Classification Monoclonal
Clone AFA-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Aflatoxin B1.
Dilution Immunoassay (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Purified aflatoxin B1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes aflatoxin B1.
Target Species Fungi
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.