Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFP, Mouse, Clone: 057-11301, Abnova™

Catalog No. 89111659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-659 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-659 Supplier Abnova Corporation Supplier No. MAB4002
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against AFP.

This gene encodes alpha-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. [provided by RefSeq

Specifications

Antigen AFP
Applications ELISA
Classification Monoclonal
Clone 057-11301
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against AFP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide)
Gene AFP
Gene Alias FETA/HPAFP
Gene Symbols AFP
Host Species Mouse
Immunogen Human AFP.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 174
Test Specificity human Alpha Fetoprotein (AFP).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.