Learn More
AFP, Mouse, Clone: 1305, Abnova™
Mouse monoclonal antibody raised against native AFP.
Supplier: Abnova Corporation MAB4005
Description
This gene encodes alpha-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. [provided by RefSeq
Specifications
| AFP | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| AFP | |
| AFP | |
| Native purified AFP from human cord blood. | |
| RUO | |
| 174 | |
| Human | |
| Liquid |
| ELISA, Radioimmune Assays (RIA) | |
| 1305 | |
| Mouse monoclonal antibody raised against native AFP. | |
| In 15mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide) | |
| FETA/HPAFP | |
| Mouse | |
| 100 μg | |
| Primary | |
| No cross-reactivity with human Albumin, CEA, Ferritin, hCG, PSA. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.