Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGER, Mouse, Clone: YK-2B4, Abnova™

Catalog No. 89108769 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-769 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-769 Supplier Abnova Corporation Supplier No. MAB10578
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant AGER.

This gene encodes a member of the immunoglobulin superfamily of cell surface molecules. It is a receptor for various molecules, including the amyloidogenic form of serum amyloid A, amyloid-beta protein, members of the S100/calgranulin superfamily and advanced glycation end products. The gene lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen AGER
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone YK-2B4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant AGER.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM HEPES, pH 7.5, 150mM NaCl (50% glycerol, 0.1mg/mL BSA)
Gene AGER
Gene Alias MGC22357/RAGE
Gene Symbols AGER
Host Species Mouse
Immunogen Recombinant protein corresponding to excellular domain of human AGER.
Purification Method Protein A-Sepharose chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 177
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.