Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGR2 and AGR3, Mouse, Clone: AGR3.4, Abnova™

Catalog No. 89136221 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-221 Supplier Abnova Corporation Supplier No. MAB6454
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against AGR2 and AGR3.

Specifications

Antigen AGR2 and AGR3
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AGR3.4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against AGR2 and AGR3.
Dilution Immunocytochemistry (1 ug/mL) Immunohistochemistry (5 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Purified human AGR2 and AGR3 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the PLMII epitope of AGR3 (AG3) and AGR2 (AG2) proteins (19-20 KDa); secreted cytoplasmic proteins which can serve as markers of carcinogenesis.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.