Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGR2, Mouse, Clone: 7E9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant AGR2.
Supplier: Abnova Corporation H00010551M04A
Description
Sequence: MEKIPVSAFLLLVALSYTLARDTTVKPGAKKDTKDSRPKLPQTLSRGWGDQLIWTQTYEEALYKSKTSNKPLMIIHHLDECPHSQALKKVFAENKEIQKLAEQFVLLNLVYETTDKHLSPDGQYVPRIMFVDPSLTVRADITGRYSNRLYAYEPADTALLLDNMKKALKLLKTELSpecifications
AGR2 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC015503 | |
AGR2 | |
AGR2 (AAH15503.1,1 a.a. ∽ 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
10551 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
7E9 | |
Mouse monoclonal antibody raised against a full-length recombinant AGR2. | |
AGR2 | |
AG2/GOB-4/HAG-2/XAG-2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction