Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahi1, Mouse, Clone: 645s3, Abnova™

Catalog No. 89113659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-659 Supplier Abnova Corporation Supplier No. MAB8962
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Ahi1.

Specifications

Antigen Ahi1
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 645s3
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Ahi1.
Dilution Western Blot (1:1000-1:2000) Immunoprecipitation (1-2 ug/200-800 ug protein) IHC (1:200-1:1000) The optimal working dilution should be determined.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene Ahi1
Gene Alias 1700015F03Rik/Ahi-1/D10Bwg0629e
Gene Symbols Ahi1
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of mouse Ahi-1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52906
Test Specificity This antibody only specifically reacts with murine Ahi-1.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.