Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHNAK, Mouse, Clone: EM-09, Abnova™

Catalog No. 89111496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-496 Supplier Abnova Corporation Supplier No. MAB3607
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant AHNAK.

Specifications

Antigen AHNAK
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone EM-09
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant AHNAK.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene AHNAK
Gene Alias AHNAKRS/MGC5395
Gene Symbols AHNAK
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus human AHNAK.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79026
Test Specificity This antibody reacts with AHNAK, a 700 KDa multi-functional adaptor proteinexpressed mainly in epithelial cell, various types of muscle cells and immune cells.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.