Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105957 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-957 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-957 Supplier Abnova Corporation Supplier No. H00008852A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant AKAP4.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is localized to the sperm flagellum and may be involved in the regulation of sperm motility. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq

Sequence: MMAYSDTTMMSDDIDWLRSHRGVCKVDLYNPEGQQDQDRKVICFVDVSTLNVEDKDYKDAASSSSEGNLNLGSLEEKEIIVIKDTEKKDQSKTEGSVCLF

Specifications

Antigen AKAP4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant AKAP4.
Formulation 50% glycerol
Gene AKAP4
Gene Accession No. NM_003886
Gene Alias AKAP82/FSC1/HI/hAKAP82/p82
Gene Symbols AKAP4
Host Species Mouse
Immunogen AKAP4 (NP_003877, 1 a.a. to 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8852
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.