Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1B1, Mouse, Clone: 7, Abnova™

Catalog No. 89113943 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-943 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-943 Supplier Abnova Corporation Supplier No. MAB9985
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant AKR1B1.

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member catalyzes the reduction of a number of aldehydes, including the aldehyde form of glucose, and is thereby implicated in the development of diabetic complications by catalyzing the reduction of glucose to sorbitol. Multiple pseudogenes have been identified for this gene. The nomenclature system used by the HUGO Gene Nomenclature Committee to define human aldo-keto reductase family members is known to differ from that used by the Mouse Genome Informatics database. [provided by RefSeq

Specifications

Antigen AKR1B1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant AKR1B1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene AKR1B1
Gene Alias ADR/ALDR1/ALR2/AR/MGC1804
Gene Symbols AKR1B1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 33-316 of human AKR1B1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231
Test Specificity It can expression in Jurkat whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.