Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1, Mouse, Clone: LP18., Abnova™

Catalog No. 89113733 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-733 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-733 Supplier Abnova Corporation Supplier No. MAB9711

Mouse monoclonal antibody raised against synthetic phosphopeptide of AKT1.

The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq

Specifications

Antigen AKT1
Applications Immunohistochemistry
Classification Monoclonal
Clone LP18.
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of AKT1.
Dilution Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Gene AKT1
Gene Alias AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA
Gene Symbols AKT1
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S473 of human AKT1.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.