Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKTide-2T, Mouse, Clone: AT-3E2, Abnova™

Catalog No. 89134564 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-564 Supplier Abnova Corporation Supplier No. MAB0014

Mouse monoclonal antibody raised against synthetic phosphopeptide AKTide-2T.

Specifications

Antigen AKTide-2T
Applications ELISA
Classification Monoclonal
Clone AT-3E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide AKTide-2T.
Dilution ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES/KOH, 150mM NaCl, pH 7.5 (50% glycerol)
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to residues ARKRERTY(pS)FGHHA, which is synthetic substrate for AKTs of AKTide-2T.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.