Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALB, Mouse, Clone: 2A3E6, Abnova™

Catalog No. 89113419 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-419 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-419 Supplier Abnova Corporation Supplier No. MAB8295
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ALB.

Specifications

Antigen ALB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A3E6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ALB.
Dilution Western Blot (1:300-1:1000) ELISA (1: 10000) The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene ALB
Gene Alias MGC179277
Gene Symbols ALB
Host Species Mouse
Immunogen Native purified ALB from bovine serum albumin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280717
Target Species Bovine
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.