Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALB, Mouse, Clone: AL-01, Abnova™

Catalog No. 89111493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-493 Supplier Abnova Corporation Supplier No. MAB3598
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ALB.

Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin. [provided by RefSeq

Specifications

Antigen ALB
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone AL-01
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ALB.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ALB
Gene Alias DKFZp779N1935/PRO0883/PRO0903/PRO1341
Gene Symbols ALB
Host Species Mouse
Immunogen Native purified human ALB.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity This antibody reacts only with human serum albumin, a 65-67 KDa monomeric proteinin human blood plasma; it is produced in liver.No cross-reactivity was observed with other serum proteins.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.