Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALCAM, Mouse, Clone: L50, Abnova™

Catalog No. 89111057 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-057 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-057 Supplier Abnova Corporation Supplier No. MAB2533

Mouse monoclonal antibody raised against ALCAM.

Specifications

Antigen ALCAM
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone L50
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against ALCAM.
Dilution Western Blot (5 ug/mL ) Immunohistochemistry (Frozen sections) (5 ug/mL) Flow Cytometry (5 ug/mL for staining 105 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene ALCAM
Gene Alias CD166/FLJ38514/MEMD/MGC71733
Gene Symbols ALCAM
Host Species Mouse
Immunogen Human ALCAM.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214
Test Specificity The monoclonal is reactive with Fc-ALCAM on Western blot. It blocks ALCAM-ALCAM interations and is reactive with the C-type domains.This antibody recognizes an epitope in two domains close to the membrane.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type ALCAM monoclonal antibody, clone L50
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.