Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDOB, Mouse, Clone: 10, Abnova™

Catalog No. 89113907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-907 Supplier Abnova Corporation Supplier No. MAB9949
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant ALDOB.

Fructose-1,6-bisphosphate aldolase (EC 4.1.2.13) is a tetrameric glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Vertebrates have 3 aldolase isozymes which are distinguished by their electrophoretic and catalytic properties. Differences indicate that aldolases A, B, and C are distinct proteins, the products of a family of related 'housekeeping' genes exhibiting developmentally regulated expression of the different isozymes. The developing embryo produces aldolase A, which is produced in even greater amounts in adult muscle where it can be as much as 5% of total cellular protein. In adult liver, kidney and intestine, aldolase A expression is repressed and aldolase B is produced. In brain and other nervous tissue, aldolase A and C are expressed about equally. There is a high degree of homology between aldolase A and C. Defects in ALDOB cause hereditary fructose intolerance. [provided by RefSeq]

Specifications

Antigen ALDOB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant ALDOB.
Dilution Western blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene ALDOB
Gene Accession No. NM_000035
Gene Symbols ALDOB
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human ALDOB.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229
Test Specificity It can expression in HEK293 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.