Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-alpha-Fetoprotein/AFP, Clone: D10, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450120.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human, Bovine (Negative), Canine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
alpha-Fetoprotein/AFP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
P02771 | |
AFP | |
Alpha fetoprotein (AFP) purified from serum of a hepatoma patient. | |
Protein A or G purified | |
RUO | |
Primary | |
It recognizes an oncofetal glycoprotein with a single chain of 70kDa, which is identified as alpha-fetoprotein (AFP) (ISOBM TD-2 workshop and assigned by K. Nustad to group D of a cluster of 6 major epitopes of human alpha fetoprotein). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors. | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
D10 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
Mouse | |
70 kDa | |
0.2 mg | |
Cancer, Cancer Stem Cells, GPCR, Stem Cell Markers, Tumor Biomarkers | |
174 | |
Human, Bovine (Negative), Canine (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction