Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-alpha-Synuclein (aa 121-125), (Ascites Free), Clone: Syn211,

Catalog No. 36008CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-008-CMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-008-CMI Supplier MilliporeSigma™ Supplier No. 36008C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects alpha-Synuclein (aa 121-125), (Ascites Free) clone: Syn211 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

Alpha-synuclein is encoded by the SNCA gene (also known as NACP, PARK1) in human. There are two other known synuclein family members, beta- and gamma-synuclein. Interest in synuclein research started with the discovery of alpha-synuclein mutation in several families with autosomal dominant Parkinson's disease (PD). Alpha-synuclein is abundantly expressed in the brain and is found in a classic amyloid fibril form within the intra-neuronal Lewy body deposits of PD. The physiological function of synucleins is not well understood, but appears to involve membrane interactions, and in particular reversible binding to synaptic vesicle membranes. Contradictory evidences regarding inhibitory action of alpha-synuclein against phospholipase D (PLD) have been reported (PMID 11821392 & 19146388). Clone Syn211, detects epitope in the region from amino acid 121 to 125 and has been used together with clone LB509, to characterize the digestion pattern of different forms of in vitro prepared alpha-synclulein aggregates (Guo, J.L., et al. 2013; PMID 23827677).
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Synuclein (aa 121-125), (Ascites Free)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Syn211
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P37840
Gene Symbols SNCA; NACP; PARK1
Host Species Mouse
Immunogen Conjugated recombinant protein corresponding to human alpha-Synuclein.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000336
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.