Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR, Mouse, Clone: 2A10F3, Abnova™

Catalog No. 89108630 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-630 Supplier Abnova Corporation Supplier No. MAB10444
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant AMACR.

Alpha-methylacyl-CoA racemase is responsible for the conversion of pristanoyl-CoA and C27-bile acyl-CoAs to their (S)-stereoisomers, which are the only stereoisomers that can be degraded via peroxisomal beta-oxidation.[supplied by OMIM

Specifications

Antigen AMACR
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2A10F3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant AMACR.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascitic (0.03% sodium azide)
Gene AMACR
Gene Alias RACE
Gene Symbols AMACR
Host Species Mouse
Immunogen Recombinant protein corresponding to human AMACR.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23600
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.